Skip to Content
Merck
All Photos(3)

Key Documents

L5908

Sigma-Aldrich

Lacto-N-fucopentaose I

from human milk, ≥90% (HPAE/PAD)

Synonym(s):

α-L-Fuc (1→2)-β-D-Gal-(1→3)-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-D-Glc, LNFP I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H55NO25
CAS Number:
Molecular Weight:
853.77
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

human milk

Assay

≥90% (HPAE/PAD)

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@H]3[C@H](O)[C@@H](CO)O[C@@H](O[C@H]4[C@@H](O)[C@@H](CO)O[C@@H](O[C@H]([C@H](O)CO)[C@H](O)[C@@H](O)C=O)[C@@H]4O)[C@@H]3NC(C)=O)[C@@H](O)[C@H](O)[C@@H]1O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Blood group H-active pentasaccharide.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Lindenberg
Danish medical bulletin, 38(5), 371-380 (1991-10-01)
6.1. Attachment and penetration of the uterine epithelium in the human: The present studies describe to our knowledge the only studies of human in vitro attachment of blastocysts to uterine epithelial monolayers. From these studies, using other mammalian species as
Carbohydrate binding properties of mouse embryos.
S Lindenberg et al.
Journal of reproduction and fertility, 89(2), 431-439 (1990-07-01)
Elin Säwén et al.
The journal of physical chemistry. B, 115(21), 7109-7121 (2011-05-07)
The conformational dynamics of the human milk oligosaccharide lacto-N-fucopentaose (LNF-1), α-L-Fucp-(1 → 2)-β-D-Galp-(1 → 3)-β-D-GlcpNAc-(1 → 3)-β-D-Galp-(1 → 4)-D-Glcp, has been analyzed using NMR spectroscopy and molecular dynamics (MD) computer simulations. Employing the Hadamard (13)C-excitation technique and the J-HMBC experiment
Andrew Almond et al.
Biochemistry, 43(19), 5853-5863 (2004-05-12)
Fucosylated O- and N-linked glycans are essential recognition molecules in plants and animals. To understand how they impart their functions, through interactions with proteins, requires a detailed analysis of structure and dynamics, but this is presently lacking. In this study
S Lindenberg et al.
Journal of reproduction and fertility, 83(1), 149-158 (1988-05-01)
Seven oligosaccharides isolated from human milk were tested for their effect in an in-vitro model of mouse blastocyst adhesion and trophoblast outgrowth on endometrial epithelial monolayers. One compound, lacto-N-fucopentaose I (LNF I), produced a significant reduction in the percentage of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service