Skip to Content
Merck
All Photos(1)

Key Documents

L5018

Sigma-Aldrich

Leucopyrokinin fragment 4-8

≥97% (HPLC)

Synonym(s):

Phe-Thr-Pro-Arg-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H49N9O6
CAS Number:
Molecular Weight:
631.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

[H][C@](NC(=O)[C@@H](N)Cc1ccccc1)([C@@H](C)O)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(N)=O

Biochem/physiol Actions

The C-terminal pentapeptide of leucopyrokinin, retaining 30% of the activity of the parent.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Nachman et al.
Biochemical and biophysical research communications, 137(3), 936-942 (1986-06-30)
Evaluation of analogs of the blocked insect myotropic neuropeptide leucopyrokinin (LPK) has demonstrated its relative insensitivity to amino acid substitution in the N-terminal in contrast to the C-terminal region. Truncated analogs of LPK without the first, second, and third N-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service