Skip to Content
Merck
All Photos(1)

Key Documents

K3644

Sigma-Aldrich

K 858

≥98% (HPLC)

Synonym(s):

K858, N-(4-Acetyl-4,5-dihydro-5-methyl-5-phenyl-1,3,4-thiadia zol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€260.00

€260.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€260.00

About This Item

Empirical Formula (Hill Notation):
C13H15N3O2S
CAS Number:
Molecular Weight:
277.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€260.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)NC1=NN(C(C)=O)C(C)(S1)c2ccccc2

InChI

1S/C13H15N3O2S/c1-9(17)14-12-15-16(10(2)18)13(3,19-12)11-7-5-4-6-8-11/h4-8H,1-3H3,(H,14,15,17)

InChI key

JEFVYQYZCAVNTP-UHFFFAOYSA-N

Biochem/physiol Actions

A thiadiazoline derivative reported as a selective ATP-uncompetitive mitotic kinesin Eg5 inhibitor.
Eg5 is a key protein involved in the formation of bipolar spindles. K858 is a selective ATP-uncompetitive mitotic kinesin Eg5 inhibitor. The compound induces mitotic arrest, caspase-3 activation and cell growth inhibition in HCT116 cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service