Skip to Content
Merck

I6277

Anti-Interleukin-2 Soluble Receptor β antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-2 sRβ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), Neutral, WB
Citations:
5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable, neutralization: suitable, western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL2RB(3560)

General description

Interleukin-2 (IL-2) is a cytokine produced by activated T cells in response to antigen stimulation. IL-2 is important for proliferation of lymphocytes, natural killer cells and macrophages and in the differentiation of CD4+ cells into Th1 and Th2 effector cells. The effects of IL-2 are mediated by the signalling activated by its binding to IL-2 receptor (IL-2 R). IL-2 R which contains three membrane subunits: α, β and γ. The β and the γ subunits complex results in the binding of IL-2 with higher affinity. IL-2 R β is expressed on resting monocytes and B cells, CD8+ and CD4+T cells, NK cells and neutrophils and mature thymocytes. IL-2 R activates key signalling pathways such as MAPK, JAK/STAT as well as PI3K/Akt. Activation of these pathways modulates production of inflammatory cytokines, survival and progression through cell cycle
Anti-Human IL-2 soluble receptor β (IL-2 sRβ) specifically reacts with human IL-2 Rβ. The antibody shows less than 15% cross reactivity with recombinant human IL-3 sRα, less than 5% cross reactivity with recombinant human IL-1 RII, IL-2 sRγ, IL-5 sRα, and less than 2% cross reactivity with recombinant human IL-2 sRα, IL-4 sR, and IL-5sRβ. In addition, the antibody shows no cross reactivity with other cytokines tested.

Immunogen

recombinant human IL-2 soluble receptor β (IL-2 sRβ) expressed in Sf 21 insect cells.

Application

Anti-Human IL-2 soluble receptor β (IL-2 sRβ) antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For indirect ELISA, the recommended concentration is 0.5-1.0 μg/ml. The antibody is suitable for IL-2 sRβ neutralization reactions at a working antibody concentration of 10-30 μg/ml.

Biochem/physiol Actions

The antibody will neutralize human cell surface IL-2 receptor β mediated-bioactivity. For cells expressing high affinity IL-2 receptors, use of both anti-IL-2 Rα and anti-IL-2 Rβ is recommended. The antibody shows ≤15% cross-reactivity with recombinant human IL-3 sRα, ≤5% cross-reactivity with recombinant human IL-1 RII, IL-2 sRγ, IL-5 sRα, and ≤2% cross-reactivity with recombinant human IL-2 sRα, IL-4 sR, and IL-5 sRβ.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library