Skip to Content
Merck
All Photos(1)

Key Documents

I5886

Sigma-Aldrich

L-Isoleucinamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O · HCl
CAS Number:
Molecular Weight:
166.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.CC[C@H](C)[C@H](N)C(N)=O

InChI

1S/C6H14N2O.ClH/c1-3-4(2)5(7)6(8)9;/h4-5H,3,7H2,1-2H3,(H2,8,9);1H/t4-,5-;/m0./s1

InChI key

SAOZPTBFWAEJQL-FHAQVOQBSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service