Skip to Content
Merck
All Photos(1)

Documents

HPA006914

Sigma-Aldrich

Anti-RPA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RF-A, Anti-RP-A, Anti-Replication factor A protein 1, Anti-Replication protein A 70 kDa DNA-binding subunit, Anti-Single-stranded DNA-binding protein, Anti-p70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SLSHTSGGTQSKVVPIASLTPYQSKWTICARVTNKSQIRTWSNSRGEGKLFSLELVDESGEIRATAFNEQVDKFFPLIEVNKVYYFSKGTLKIANKQFTAVKNDYEMTFNNETSVMPCEDDHHLPTVQFDFTGIDD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA1(6117)

Immunogen

Replication protein A 70 kDa DNA-binding subunit recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPA1 (Replication protein A1) is the largest component (70kDa) of the heterotrimeric complex RPA with three functional domains. It plays a pivotal role in the DNA replication, repair, and recombination. It also participates in the transcriptional regulation of various genes. Its zinc finger domain binds to the single-stranded DNA to form the RPA holocomplex. In addition, it also helps to unwind the double strand, stimulates DNA polymerases α and δ, and form the initial short Okazaki fragments. At the damage site, RPA1 recruits DNA double-strand break repair factors, RAD51 and RAD52 to the chromatin. RPA1 plays an essential role in the DNA replication and mismatch repair but it is not mandatory for nucleotide excision repair. In the nucleotide excision repair, it recruits DNA damage proteins like XPA and XPG.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71063

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y L Lin et al.
The Journal of biological chemistry, 273(3), 1453-1461 (1998-01-27)
The largest subunit of the replication protein A (RPA) contains an evolutionarily conserved zinc finger motif that lies outside of the domains required for binding to single-stranded DNA or forming the RPA holocomplex. In previous studies, we showed that a
E van der Knaap et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(18), 9979-9983 (1997-09-02)
Internodes of deepwater rice are induced to grow rapidly when plants become submerged. This adaptation enables deepwater rice to keep part of its foliage above the rising flood waters during the monsoon season and to avoid drowning. This growth response
A Aboussekhra et al.
Cell, 80(6), 859-868 (1995-03-24)
Nucleotide excision repair is the principal way by which human cells remove UV damage from DNA. Human cell extracts were fractionated to locate active components, including xeroderma pigmentosum (XP) and ERCC factors. The incision reaction was then reconstituted with the
Kate M Sleeth et al.
Journal of molecular biology, 373(1), 38-47 (2007-09-04)
The replication protein A (RPA) is involved in most, if not all, nuclear metabolism involving single-stranded DNA. Here, we show that RPA is involved in genome maintenance at stalled replication forks by the homologous recombination repair system in humans. Depletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service