Skip to Content
Merck
All Photos(2)

Documents

HPA003146

Sigma-Aldrich

Anti-ZNF44 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GIOT-2 antibody produced in rabbit, Anti-Gonadotropin-inducible transcription repressor 2 antibody produced in rabbit, Anti-Zinc finger protein 44 antibody produced in rabbit, Anti-Zinc finger protein KOX7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HEEWALLGPSQKNLYRDVMRETIRNLNCIGMKWENQNIDDQHQNLRRNPRCDVVERFGKSKDGSQCGETLSQIRNSIVNKNTPARVDACGSSVNGEVIMGHSSLNCYIRVDTGHKHRECHEYAEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF44(51710)

Related Categories

General description

The zinc finger proteins (ZNFs) are made up of anti-parallel hairpin motif. These proteins contain an α-helix, two β-strands and a hairpin structure. The ZNFs are mainly involved in the recognition of DNA, folding and assembly of proteins, transcriptional activation, lipid binding, regulation of apoptosis and packaging of RNA.

Immunogen

Zinc finger protein 44 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF44 (zinc finger protein 44) gene encodes a zinc finger protein that may function as a transcriptional regulator. The gene is localized to chromosome 19. It has been implicated in breast cancer. Deletions/defects in this gene may cause generalized epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83538

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander G Bassuk et al.
American journal of medical genetics. Part A, 161A(7), 1722-1725 (2013-05-21)
Rare copy number variants (CNVs) have been established as an important cause of various neurodevelopmental disorders, including intellectual disability (ID) and epilepsy. In some cases, a second CNV may contribute to a more severe clinical presentation. Here we present two
Identification and Analysis of Disease Target Network of Human MicroRNA and Predicting Promising Leads for ZNF439, a Potential Target for Breast Cancer.
Raghavan V and Manasa D B
International Journal of Bioscience, Biochemistry and Bioinformatics, 2, 358-362 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service