Skip to Content
Merck
All Photos(1)

Key Documents

H5533

Sigma-Aldrich

Hemin

from bovine, ≥80%

Synonym(s):

Chloro(protoporphyrinato)iron(III), Chlorohemin, Chloroprotoporphyrin IX iron(III), Ferriprotoporphyrin IX chloride, Hemin(chloride)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32ClFeN4O4
CAS Number:
Molecular Weight:
651.94
Beilstein:
5229914
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

biological source

bovine

Assay

≥80%

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3c(CCC(O)=O)c(C)c4cc5nc(cc6c(C)c(C=C)c(cc1n2)n6[Fe](Cl)n34)c(C=C)c5C

InChI

1S/C34H34N4O4.ClH.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);1H;/q;;+3/p-3/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

BTIJJDXEELBZFS-HXFTUNQESA-K

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Takanori Uzawa et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(37), 13859-13864 (2008-09-10)
The earliest steps in the folding of proteins are complete on an extremely rapid time scale that is difficult to access experimentally. We have used rapid-mixing quench-flow methods to extend the time resolution of folding studies on apomyoglobin and elucidate
Bingying Jiang et al.
Biosensors & bioelectronics, 43, 289-292 (2013-01-22)
In this work, we describe the development of a sensitive and label-free aptasensor for thrombin detection based on background noise reduction by exonuclease I (Exo I) and signal amplification by direct electron transfer (DET) of hemin. The thrombin binding aptamers
Dipankar Sen et al.
Critical reviews in biochemistry and molecular biology, 46(6), 478-492 (2011-10-01)
Guanine-rich RNAs and DNAs from chromosomal telomeres and elsewhere that fold into guanine quadruplexes (G-quadruplexes), are found to complex tightly with porphyrins such as N-methylmesoporphyrin IX (NMM) and hemin [Fe(III) heme]. By themselves, these DNAs and RNAs are found to
V Herbert et al.
Stem cells (Dayton, Ohio), 12(3), 289-303 (1994-05-01)
Iron is a double-edged sword. In moderate quantities and leashed to protein, it is an essential element in all cell metabolism and growth, but it is toxic when unleashed. Because of its ability to switch back and forth between ferrous
Stephen R Robinson et al.
Redox report : communications in free radical research, 14(6), 228-235 (2009-12-17)
Hemorrhagic stroke is a common cause of permanent brain damage, with a significant amount of the damage occurring in the weeks following a stroke. This secondary damage is partly due to the toxic effects of hemin, a breakdown product of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service