Skip to Content
Merck
All Photos(1)

Key Documents

H2505

Supelco

Hexyl nicotinate

Synonym(s):

Nicotinic acid hexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

density

1.02 g/mL at 25 °C (lit.)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service