Skip to Content
Merck
All Photos(3)

Key Documents

H1916

Sigma-Aldrich

Anti-HOXC9 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HOX3B, Anti-Homeobox 3B, Anti-Homeobox C9, Anti-Hox-3.2 (MOUSE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2-4 μg using lysates of HEK-293T cells over expressing human HOXC9
indirect immunofluorescence: 1-2 μg/mL using paraformaldehyde fixed HEK-293T cells, over expressing human HOXC9
western blot: 0.5-1 μg/mL using lysates of HEK-293T cells over expressing human HOXC9

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXC8(3224)
mouse ... Hoxc8(15426)
rat ... Hoxc9(368178)

General description

Homeobox C9 (HOXC9) gene is a member of the conserved homeobox family and is located on the human chromosome at 12q13.13. This gene encodes for the HOXC9 transcription factor/protein.

Specificity

Anti-HOXC9 (C-terminal) recognizes human HOXC9 (also known as Hox-3B).

Application

Anti-HOXC9 (C-terminal) antibody produced in rabbit may be used in
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Homeobox C9 (HOXC9) transcription factor/ protein is involved in morphogenesis. Loss of this gene is associated with a hypermethylated state of its promoter in non-small cell lung cancer and breast cancer. HOXC9 protein produces neuronal differentiation and growth arrest in neuroblastoma cells and acts as a biomarker in neuroblastoma patients. HOXC9 gene expression is upregulated by retinoic acid. The HOXC9 gene expression is associated with astrocytomas malignancy and cervical cancer.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R López et al.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 16(3), 1289-1296 (2006-06-29)
The HOX genes are a family of transcription factors that bind to specific sequences of DNA in target genes regulating their expression. The role of HOX genes in adult cell differentiation is still obscure, but growing evidence suggests that they
Xudong Peng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 49(4), 1659-1676 (2018-09-12)
Previous studies demonstrated that HOXC9 acts as an oncogene in several tumors. The aim of this study was to explore whether HOXC9 promotes gastric cancer (GC) progression and elucidate the underlying molecular mechanisms. HOXC9 expression in GC tissues and adjacent
Oswaldo K Okamoto et al.
Biochimica et biophysica acta, 1769(7-8), 437-442 (2007-06-26)
Comparative analysis of cancer stem cells with their neoplastic and non-neoplastic counterparts should help better understand the underlying molecular events leading to transformation and tumor dissemination. Here, we report a molecular signature comprised by genes with exclusive aberrant expression in
Ling Mao et al.
Cancer research, 71(12), 4314-4324 (2011-04-22)
Differentiation status in neuroblastoma strongly affects clinical outcomes and inducing differentiation is a treatment strategy in this disease. However, the molecular mechanisms that control neuroblastoma differentiation are not well understood. Here, we show that high-level HOXC9 expression is associated with
Lei Lv et al.
Cancer letters, 357(1), 105-113 (2014-12-03)
Chemoresistance prevents the curative cancer chemotherapy and presents a formidable challenge for both cancer researchers and clinicians. We have previously shown that miR-193a-3p promotes the multi-chemoresistance of bladder cancer cells via repressing its three target genes: SRSF2, PLAU and HIC2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service