Skip to Content
Merck
All Photos(1)

Documents

H0916

Sigma-Aldrich

Anti-HOXA10 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HOMEOBOX 1H, Anti-HOMEOBOX A10, Anti-HOX-1.8, Anti-HOX1H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using lysates of HEK-293T cells over expressing human HOXA10

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXA10(3206)

General description

The homeobox A10 (HOXA10) protein is localized in the adult human endometrium. The HOXA10 gene is mapped on the human chromosome at 7p15.2.

Specificity

Anti-HOXA10 (N-terminal) recognizes human HOXA10 (also known as Hox-1H and HOX-1).

Application

Anti-HOXA10 (N-terminal) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Homeobox A10 (HOXA10) protein is critical for the normal development of the erythroid and megakaryocytic lineages. The expression of the HOXA10 gene was found to be altered in endometriosis. HOXA10 protein plays a role in mediating endometrial development during the menstrual cycle. This protein is also involved in preparing the endometrium for implantation in humans.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Julie Kim et al.
Molecular human reproduction, 13(5), 323-332 (2007-03-14)
Endometriosis is a poorly understood gynaecologic disorder that is associated with infertility. In this study, we examined the expression of HOXA10 in the eutopic endometrium of baboons with induced endometriosis. A decrease in HOXA10 mRNA was observed after 3, 6
H S Taylor et al.
The Journal of clinical investigation, 101(7), 1379-1384 (1998-04-29)
Hox genes are well-known transcriptional regulators that play an essential role in directing embryonic development. Mice that are homozygous for a targeted disruption of the Hoxa10 gene exhibit uterine factor infertility. We have recently demonstrated that HOXA10 is expressed in
Alysson Zanatta et al.
Journal of assisted reproduction and genetics, 27(12), 701-710 (2010-09-08)
endometriosis and its associated infertility have been the object of continuous research for over a century. To understand the molecular mechanisms underlying the disease, it has become necessary to determine the aspects of its etiology that are not explained by
Nan Zhu et al.
The Journal of clinical investigation, 126(3), 997-1011 (2016-02-16)
Self-renewal is a hallmark of both hematopoietic stem cells (HSCs) and leukemia stem cells (LSCs); therefore, the identification of mechanisms that are required for LSC, but not HSC, function could provide therapeutic opportunities that are more effective and less toxic
Edward J Richards et al.
Oncotarget, 6(33), 34745-34757 (2015-10-03)
The homeobox A (HOXA) region of protein-coding genes impacts female reproductive system embryogenesis and ovarian carcinogenesis. The 5-prime end of HOXA includes three long non-coding RNAs (lncRNAs) (HOXA10-AS, HOXA11-AS, and HOTTIP) that are underexplored in epithelial ovarian cancer (EOC). We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service