Skip to Content
Merck
All Photos(1)

Documents

GW21046

Sigma-Aldrich

Anti-CCR2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Chemokine (C-C motif) receptor 2 isoform A, Anti-MCP-1 receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21046

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCR2(1231)

General description

CCR2 (chemokine (C-C motif) receptor 2) belongs to the chemokine receptor family, which are small (8-10kDa) G-protein coupled receptors (GPCR). This family is subdivided into four families called CC, CXC, CX3C and XC. CCR2 belongs to the CC family, and has its ligand-binding domain in its exoplasmic N-terminal. Its C-terminal is involved in intracellular signaling. It is constitutively expressed by monocytes, and is found on T-cells, naïve B-cells, NK (natural killer) cells, dendritic cells and basophils.

Immunogen

Immunogen Sequence: GI # 4502633, sequence 1-45
Recombinant chemokine (C-C motif) receptor 2 isoform A

Application

Anti-CCR2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
.

Biochem/physiol Actions

C-C chemokine receptor type 2 (CCR2) is an important chemokine for monocyte aggregation and transendothelial monocyte migration. CCR2 plays an important role in extravasation and transmigration of monocytes under inflammatory conditions. It is involved in the growth and invasion of prostate cancer (PCa). CCR2 expression is very important for CCL2-induced proliferation and invasion of PC-3M cells. CCR2 can also act as a promising molecular target for prevention of PCa growth and metastasis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alireza Zandifar et al.
TheScientificWorldJournal, 2013, 836309-836309 (2014-01-24)
Migraine is a multifactorial common neurovascular disease with a polygenic inheritance. Inflammation plays an important part in migraine pathophysiology. C-C chemokine receptor 2 (CCR2) is an important chemokine for monocyte aggregation and transendothelial monocyte migration. The aim of our study
Hannah X Chu et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(9), 1425-1429 (2014-07-06)
CC chemokine receptor 2 (CCR2) plays important roles in extravasation and transmigration of monocytes under inflammatory conditions. CCR2 and its ligands have been extensively studied in a range of inflammatory diseases in the central nervous system (CNS), including multiple sclerosis
Ji Gao et al.
Oncology research, 21(2), 73-82 (2014-01-11)
Prostate cancer (PCa) is the second most lethal malignancy in men. It has been reported that chemokines, produced by cancer cells, have linked with tumor progression and metastatic spread. We have proven that chemokine (C-C) motif ligand 2 (CCL2) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service