Key Documents
GN791840
Greiner 3-D Aldehyde HTA plate
non-sterile, 96 tests per plate
About This Item
UNSPSC Code:
41121800
size:
96 wells
Pricing and availability is not currently available.
packaging
pack of 5 ea
manufacturer/tradename
Greiner 791840
availability
available only in China
size
96 wells
suitability
suitable for (cell culture)
General description
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA Plate has been optimized for the simultaneous analysis of multiple samples.
Features and Benefits
  • low well rim of only 0.3 mm
  • low autofluorescence
Legal Information
HTA is a trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)