Skip to Content
Merck
All Photos(1)

Key Documents

GERPN1004

Rabbit Ig

Cytiva RPN1004

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€526.00

€526.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€526.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€526.00


Please contact Customer Service for Availability

packaging

pkg of 2 mL

manufacturer/tradename

Cytiva RPN1004

storage temp.

2-8°C

Related Categories

General description

The antibodies are supplied with dilution guidelines for relevant applications.

Application

Immunodetection ELISA tests In situ hybridization Biotinylated antibodies are now used in many ways, including quantitative immunoassay, screening procedures for the detection of hybridoma antibodies, immunocytochemistry and for antigen identification in protein blotting ELISA, immunocytochemistry.[1]
The antibodies are supplied with dilution guidelines for relevant applications and are stored at 2°C to 8°C.

Features and Benefits

  • Highly species-specific.
  • Applications: Immunodetection, ELISA tests, in situ hybridization.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prenatal immune challenge disrupts sensorimotor gating in adult rats: implications for the etiopathogenesis of schizophrenia.
Borrell J, et al.
Neuropsychopharmacology, 26(2), 204-204 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service