Skip to Content
Merck
All Photos(1)

Documents

GE27-9502-01

Ready-to-go Rapid Analysis Kit

Cytiva 27-9502-01, sufficient for 100 reactions, pack of 6 primers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55101520
NACRES:
NA.55

form

solid

usage

sufficient for 100 reactions

feature

hotstart: no

packaging

pack of 6 primers

manufacturer/tradename

Cytiva 27-9502-01

color

Colorless

solubility

cold water: soluble (Soluble in cold water and hot water)

Related Categories

General description

RAPD analysis is a technique for rapidly detecting genomic polymorphisms. A single short oligonucleotide primer of arbitrary sequence is used under low stringency conditions in PCR to generate a reproducible array of strain-specific products.

Application

illustra Ready-To-Go RAPD Analysis Beads are designed as premixed , predispensed reactions for performing random amplified polymorphic DNA (RAPD) analysis. With RAPD analysis, genomic polymorphisms can be detected at multiple loci using only nanogram quantities of DNA. The RAPD reactions are provided as room-temperature-stable dried beads that contain all the necessary reagents, except primer, for performing RAPD analysis.

RAPD analysis is a technique for rapidly detecting genomic polymorphisms. A single, short oligonucleotide primer of arbitrary sequence is used under Low stringency conditions in PCR to generate a reproducible array of strain-specific products that are analyzed by gel electrophoresis. Under these conditions, genomic polymorphisms can be detected at multiple loci using only nanogram quantities of DNA. RAPD analysis has been used in numerous applications, including gene mapping, detection of strain diversity, population analysis, epidemiology and the analysis of phylogenetic and taxonomic relationships (2).


The RAPD reactions are provided as dried beads that contain all the necessary PCR components, except primer, in concentrations optimized for RAPD analysis. Beads are provided in thin-walled 0.5 mL tubes compatible with most thermal cyclers. Each package of Ready-To-Go RAPD Analysis Beads contains sufficient reagents for 100 individual RAPD reactions: RAPD analysis beads, control E. coli BL21 DNA, control E. coli C1a DNA, RAPD analysis primer 2, and instruction booklet (DNA from two E. coli strains and RAPD analysis primer 2 are provided as controls to assay the ability of the RAPD beads to amplify DNA and identify polymorphisms).

Features and Benefits

  • For performing DNA profiling experiments using the randomly amplified polymorphic DNA [RAPD (1)] technique.
  • Preformulated, predispensed, single-dose reaction beads are provided as ambient-temperature-stable beads to ensure greater reproducibility between reactions, minimize pipetting steps and reduce the potential for pipetting errors and contamination.
  • RAPD* reactions are simple to perform—simply add genomic DNA solution and primer to a tube of RAPD Analysis Beads to a final reaction volume of 25 μL and cycle the reaction.
  • Each lot of RAPD Analysis Beads is function-tested to ensure its ability to generate a differential banding pattern between the two control E. coli strains using RAPD Analysis Primer 2 included with the RAPD beads.
  • RAPD reactions are pre-optimized for use with a wide variety of organisms.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ready-To-Go is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service