Skip to Content
Merck
All Photos(1)

Documents

GE27-5140-01

Microspin S-400 HR

Cytiva 27-5140-01, pack of 50 columns

Synonym(s):

Microspin S-400 HR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

packaging

pack of 50 columns

manufacturer/tradename

Cytiva 27-5140-01

storage condition

(20% Ethanol)
(TE buffer (10 mM Tris/HCl, 1mM EDTA, pH 7.6))

matrix

allyl dextran and N, N’-methylenebisacrylamide

particle size

25-75 μm

avg. part. size

50 μm

cleaning

2-13

working range

3-11

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

MicroSpin columns are designed for the rapid purification of nucleic acids for use in a wide range of applications, including desalting, buffer exchange and removal of primers. Good product yield and purity is obtained with sample volumes from 25-100 μl, and from nanogram to milligram quantities of DNA.

Features and Benefits

  • illustra MicroSpin S-400 HR Columns: For rapid purification of PCR products (>200 bp) from unincorporated primers (<32-mers) and nucleotides using spin-column chromatography. - Accommodates 25 to 50 μ for post-PCR cleanup prior to cloning or a second amplification reaction or 51 to 100 μ for all other applications.
  • Convenient: Prepacked with Sephacryl® S-400 HR pre-equilibriated in TE buffer.
  • Ready to use: Requires less than 4 min from sample application to collection of purified product.
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

MicroSpin is a trademark of Cytiva
Sephacryl is a registered trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service