Key Documents
GE18112090
Union 1/16" Male - 1/16" Male
size (0.13 mm), ÄKTA® Systems, Cytiva 18112090
Select a Size
1 EACH
€163.00
€163.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€163.00
About This Item
UNSPSC Code:
41116500
NACRES:
NB.13
€163.00
Please contact Customer Service for Availability
material
PEEK
feature
Connections: Fingertight 1/16"
Wetted Material: Polyetheretherketone (PEEK)
ÄKTA® Systems
manufacturer/tradename
Cytiva 18112090
size
(0.13 mm)
compatibility
Tubing o.d. 1/16"
Legal Information
ÄKTA is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)