Skip to Content
Merck
All Photos(1)

Key Documents

G9916

SAFC

EX-CELL® GTM-3

Animal-component free, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Gene Therapy Medium-3 for Adenovirus Production

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€162.00
6 X 1 L
€668.00

€162.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
€162.00
6 X 1 L
€668.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

€162.00


Please contact Customer Service for Availability

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: 2.93 g/L

shipped in

ambient

storage temp.

2-8°C

Features and Benefits

Complete, ready-to-use medium that requires only the addition of L-glutamine for the growth of HEK-293 cells, retinoblastoma-like cells and propagation of adenoviruses. Additionally, the medium is designed to support growth of cells in suspension culture with minimal levels of cell clumping. Developed to meet the needs of biotechnology and vaccine manufacturing.

Caution

Do not freeze.

Reconstitution

Prior to use medium should be warmed to 37°C and supplemented with 20ml/L of 200mM L-glutamine.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Berdichevsky et al.
Biotechnology progress, 24(1), 158-165 (2007-12-07)
PER.C6 cells, an industrially relevant cell line for adenovirus manufacture, were extensively passaged in serum-free suspension cell culture to better adapt them to process conditions. The changes in cell physiology that occurred during this passaging were characterized by investigating cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service