Skip to Content
Merck
All Photos(1)

Documents

G9173

Sigma-Aldrich

β2-Glycoprotein I from human plasma

Synonym(s):

Apolipoprotein H

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

human plasma

Quality Level

Assay

≥96% (SDS-GE)

form

lyophilized

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APOH(350)

Related Categories

Application

Possible target of antiphospholipid antibodies and a component involved in phospholipid immunogenicity in the APA syndrome.

Physical form

Purified protein containing glycine and NaCl as stabilizers

Preparation Note

Store reconstituted product at 4°C no longer than 4 weeks.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service