Skip to Content
Merck
All Photos(1)

Key Documents

G7160

Sigma-Aldrich

Galanin Fragment 1-13-Spantide I

≥95% (HPLC)

Synonym(s):

C7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C138H199N35O30
CAS Number:
Molecular Weight:
2828.27
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

Gene Information

Amino Acid Sequence

Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-D-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2

Biochem/physiol Actions

Tandem-linked peptide which acts as a potent galanin receptor antagonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L G Ulman et al.
Regulatory peptides, 51(1), 17-23 (1994-04-14)
Previous studies have shown that injection of galanin (GAL: 6.2 nmol/kg) causes prolonged inhibition of cardiac vagal action in anaesthetised cats. Stimulation of the cardiac sympathetic nerve (16 Hz for 5 min) also produces inhibition of cardiac vagal action, an
I Kisfalvi et al.
Journal of physiology, Paris, 94(1), 37-42 (2000-04-13)
The neuropeptide galanin has been reported to have a wide range of biological actions both in the central nervous system and in the gastrointestinal tract. Recent works led to the discovery of selective galanin receptor antagonists including M15 (galanin(1-12)-Pro-substanceP(5-11)-amide), M35
D A Mahns et al.
Regulatory peptides, 67(3), 163-168 (1996-12-17)
Galanin is a neuropeptide that causes a marked pressor response in several non-mammalian vertebrate species, and some marsupials. In this study, the effect of three galanin antagonists were tested on the pressor response to an intravenous dose (6.3 nmol/kg) of
I Kisfalvi et al.
Journal of physiology, Paris, 95(1-6), 385-389 (2001-10-12)
Galanin is a neuropeptide having a wide range of biological actions. Recently selective galanin receptor antagonists such as M35 [galanin(1-12)-Pro-bradykinin(2-9)-amide] and C7 [galanin(1-12)-Pro-spantide-amide] have been described. These antagonists have blocked the actions of galanin on flexor reflex, glucose-induced insulin secretion
R L Corwin et al.
The European journal of neuroscience, 5(11), 1528-1533 (1993-11-01)
Galanin significantly increased food intake when microinjected into the region of the central nucleus of the amygdala as well as into the paraventricular nucleus of the hypothalamus. In the amygdala this effect was specific to feeding; no change in grooming

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service