Skip to Content
Merck
All Photos(1)

Key Documents

G1027

Sigma-Aldrich

Ginsenoside-Re from Panax ginseng (Korean ginseng) root

triterpenoid saponin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant root (Panax ginseng)

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2O[C@H]3C[C@]4(C)[C@H](C[C@@H](O)[C@@H]5[C@H](CC[C@@]45C)[C@](C)(CC\C=C(/C)C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@@]7(C)CC[C@H](O)C(C)(C)[C@H]37)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C48H82O18/c1-21(2)11-10-14-48(9,66-42-38(60)35(57)32(54)26(19-49)63-42)23-12-16-46(7)30(23)24(51)17-28-45(6)15-13-29(52)44(4,5)40(45)25(18-47(28,46)8)62-43-39(36(58)33(55)27(20-50)64-43)65-41-37(59)34(56)31(53)22(3)61-41/h11,22-43,49-60H,10,12-20H2,1-9H3/t22-,23-,24+,25-,26+,27+,28+,29-,30-,31-,32+,33+,34+,35-,36-,37+,38+,39+,40-,41-,42-,43+,45+,46+,47+,48-/m0/s1

InChI key

PWAOOJDMFUQOKB-WCZZMFLVSA-N

Biochem/physiol Actions

Ginsenoside Re is a phytosterol found in the ginseng root. Ginsenoside Re is believed to be the main contributor to ginseng′s protection against damage from cardiac ischemia by inhibiting calcium accumulation in the mitochondria. It has been shown that Ginsenoside Re inhibits calcium buildup through activation of endothelial nitric oxide synthase (eNOS) to release NO and activate delayed rectifier K+ currents.
Triterpene saponin found in ginseng.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginsenoside Re, a Main Phytosterol of Panax ginseng, Activates Cardiac Potassium Channels via a Nongenomic Pathway of Sex Hormones.
Furukawa, T.
Molecular Pharmacology, 6, 1916-1924 (2006)
Juan Liu et al.
Journal of experimental botany, 71(20), 6396-6407 (2020-08-15)
Adventitious root branching is vital to plant growth and regeneration, but the regulation of this process remains unclear. We therefore investigated how ginsenosides regulate adventitious root branching in Panax ginseng. Cell proliferation and adventitious root branching were decreased in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service