Skip to Content
Merck
All Photos(1)

Documents

F1500

Fast Red TR Salt hemi(zinc chloride) salt

practical grade

Synonym(s):

4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6Cl2N2 · 0.5 ZnCl2
CAS Number:
Molecular Weight:
257.19
Colour Index Number:
37085
MDL number:

grade

practical grade

composition

Dye content, ~15%

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.Cc1cc(Cl)ccc1[N+]#N.Cc2cc(Cl)ccc2[N+]#N

InChI

1S/2C7H6ClN2.4ClH.Zn/c2*1-5-4-6(8)2-3-7(5)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

RIIVFDKBXXMYRL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thaqif El Khassawna et al.
Histochemistry and cell biology, 144(5), 491-507 (2015-07-27)
Bone loss is a symptom related to disease and age, which reflects on bone cells and ECM. Discrepant regulation affects cell proliferation and ECM localization. Rat model of osteoporosis (OVX) was investigated against control rats (Sham) at young and old

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service