Skip to Content
Merck
All Photos(1)

Key Documents

F1168

Sigma-Aldrich

Fibroblast Growth Factor-9 human

≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

hGFG-9, FGF-9, GAF, Glial Activating Factor

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€223.30

€223.30

List price€319.00Save 30%

Please contact Customer Service for Availability


Select a Size

Change View
25 μG
€223.30

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€223.30

List price€319.00Save 30%

Please contact Customer Service for Availability

Product Name

Fibroblast Growth Factor-9 human, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

2.6 ng/mL

mol wt

~25 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF9(2254)

General description

FGF9 (fibroblast growth factor 9) is a member of the FGF family, which consists of a minimum of 24 members. This protein is highly conserved across species, and the gene is localized to human chromosome 13q11-q12. In embryos, FGF9 mRNA has a ubiquitous expression pattern. In adults, this protein has a limited range of expression in normal conditions, and expression is usually restricted to brain, kidney and uterus.[1]

Application

FGF9 (fibroblast growth factor 9) has been used for the culture of embryonic Sertoli cells obtained from E14.5 testes.[2]

Biochem/physiol Actions

FGF9 (fibroblast growth factor 9) is expressed in the epithelium and mesothelium of developing lung in the pseudoglandular stage. In developing lungs, it regulates the branching of epithelium, proliferation of mesenchymal cells, and expansion of the lungs. It is overexpressed in patients with idiopathic pulmonary fibrosis (IPF), in regions demonstrating active cell hyperplasia, metaplasia and fibrosis. Thus, it might facilitate the pathogenesis of IPF.[3] The expression of this protein is up-regulated in colon cancer, which is an outcome of hypoxia-induced translational activation.[1]
Mitogenic for glial cells, fibroblasts, and oligodendrocyte type 2 astrocyte progenitor cells; not mitogenic for endothelial cells. Like aFGF and bFGF, FGF-9 has no secretory signal sequence.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 1.25 mg bovine serum albumin.

Analysis Note

The biological activity is measured in a bioassay using BALB/3T3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsung-Ming Chen et al.
Nucleic acids research, 42(5), 2932-2944 (2013-12-18)
Human fibroblast growth factor 9 (FGF9) is a potent mitogen involved in many physiological processes. Although FGF9 messenger RNA (mRNA) is ubiquitously expressed in embryos, FGF9 protein expression is generally low and restricted to a few adult organs. Aberrant expression
Louise Willerton et al.
The International journal of developmental biology, 48(7), 637-643 (2004-10-08)
Proliferation and cord formation by embryonic Sertoli cells are pivotal events involved in testis morphogenesis. A number of growth factors have been implicated in mediating these events. However, the exact level of involvement and importance of each as yet remains
Fibroblast growth factors (FGFs).
Vainikka, S., et al. et al.
Guidebook to Cytokines and Their Receptors, 214-214 (1994)
M Miyamoto et al.
Molecular and cellular biology, 13(7), 4251-4259 (1993-07-01)
Glia-activating factor (GAF) is a novel heparin-binding growth factor purified from the culture supernatant of a human glioma cell line. It shows a spectrum of activity slightly different from those of other known growth factors. We have isolated the cDNA
Emily Coffey et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 61(9), 671-679 (2013-06-26)
The fibroblast growth factor (FGF) family of signaling ligands contributes significantly to lung development and maintenance in the adult. FGF9 is involved in control of epithelial branching and mesenchymal proliferation and expansion in developing lungs. However, its activity and expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service