EP4960800023
Eppendorf® Tube holder for Varispenser 25 mL/100 mL
tube holder, for Varispenser®25 mL/100 mL
Synonym(s):
Eppendorf® Varispenser®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
precision ± 0.1%
telescopic intake tube allows use with short or tall bottles.
Sterilization Compatibility
autoclavable compatible
feature
for Varispenser®25 mL/100 mL
manufacturer/tradename
Eppendorf® 4960800023 (Global)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Bottle-top dispensers with PTFE valve blocks and pistons and a Pt-lr spring borosilicate ball valve for high resistance to aggressive chemicals. Modular construction for simple cleaning and maintenance.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service