Key Documents
EP022639561
Buckets for Eppendorf® A-4-38 Rotor
rectangular, capacity 90 mL, pack of 1 ea
About This Item
UNSPSC Code:
41103910
To inquire about this product EP022639561, please contact your local Merck office or dealer. Contact Technical Service
material
aluminum
rectangular
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639561
capacity
90 mL
compatibility
for analyte group A-4-38
Related Categories
General description
Spare rectangular bucket 90 mL, for Rotor A-4-38
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)