Skip to Content
Merck
All Photos(1)

Documents

EP0030605578

Eppendorf® Microplate 96/U-PP, 96 wells

gray polypropylene, round bottom white wells, PCR clean, pkg of 240 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

gray polypropylene
round bottom white wells

packaging

pkg of 240 ea

manufacturer/tradename

Eppendorf® 0030605578

technique(s)

PCR: suitable

size

96 wells

Looking for similar products? Visit Product Comparison Guide

General description

Pipette, check and recover your samples easily with Eppendorf round bottom Microplates. The easy-to-read OptiTrack alphanumeric matrix label for identifying each well speeds up filling and significantly reduces the risk of pipetting errors.

Application

  • sample storage and preparationassays that require high resistance against temperature or solvents
  • active ingredient screening
  • combinatorial chemistry
  • storage of cDNA or genomic banks
  • protein analysis
  • genotyping
  • storage of siRNA or oligonucleotide libraries
  • preparation of tissue and plant lysates

Features and Benefits

  • Eppendorf Microplates consist of polypropylene wells which enhance sample visibility. In addition, polypropylene also ensures resistance towards chemicals, mechanical stressors and high temperatures.
  • A raised well rim and a smooth surface facilitates secure sealing.
  • Eppendorf Microplates can withstand centrifugal forces up to six thousand times the gravitational force.
  • The OptiTrack matrix feature enables faster well identification and reduces pipetting error.
  • An optimized RecoverMax well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • Eppendorf Microplates function efficiently over a wide temperature range, ranging from -86 °C to 100 °C.
  • These microplates can be autoclaved at 121 °C for 20 min.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service