Skip to Content
Merck
All Photos(1)

Key Documents

EMU185981

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Akap11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCTGGTGTTGTGTGGATTTACTCTCCAGCTACATTACACACTGCAGTAATACAGTTATGGCGGCTTTCCAGCCCCTCAGGAGTAGTCACCTAAAGAGCAAAGCGTCTGTCCGAAAAAGCTTCAGTGAAGATGTGTTCCGGTCTGTAAAGTCTTTATTACAGAGCGAGAAGGAGCTATGCAGTGTATCAGGAGGAGAGTGTTTAAACCAGGATGAGCACCCCCAGTTAACTGAGGTCACGTTTCTGGGCTTTAATGAAGAAACAGATGCTGCTCATATACAGGATCTAGCTGCAGTTTCATTGGAACTTCCAGATCTTCTGAATTCGCTCCATTTTTGCAGTCTAAGTGAAAATGAAATCATTTGTATGAAGGATACCAGTAAATCGTCCAATGTAAGCAGTAGTCCTCTAAATCAGAGTCATCATTCGGGAATGCTTTGTGTCATGAGAGTGTCACCTACATTACCGGGACTCAGAATTGATTTTATCTTTAGTCTCCTAAGTAAATATGCTGCTGGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon A Hinke et al.
The EMBO journal, 31(20), 3991-4004 (2012-09-04)
Endocrine release of insulin principally controls glucose homeostasis. Nutrient-induced exocytosis of insulin granules from pancreatic β-cells involves ion channels and mobilization of Ca(2+) and cyclic AMP (cAMP) signalling pathways. Whole-animal physiology, islet studies and live-β-cell imaging approaches reveal that ablation
Michele E Day et al.
The Journal of cell biology, 193(2), 347-363 (2011-04-20)
Although RII protein kinase A (PKA) regulatory subunits are constitutively localized to discrete cellular compartments through binding to A-kinase-anchoring proteins (AKAPs), RI subunits are primarily diffuse in the cytoplasm. In this paper, we report a novel AKAP-dependent localization of RIα

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service