Skip to Content
Merck
All Photos(1)

Key Documents

EMU082041

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cdh2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATCAATGGCAATCAAGTGGAGAACCCCATTGACATTGTCATCAATGTTATTGACATGAATGATAACAGACCTGAGTTTCTGCACCAGGTTTGGAATGGGTCTGTTCCAGAGGGATCAAAGCCTGGGACGTATGTGATGACGGTCACTGCCATTGATGCGGATGATCCAAATGCCCTGAATGGAATGCTGCGGTACAGGATCCTGTCCCAGGCGCCCAGCACACCTTCACCCAACATGTTTACAATCAACAATGAGACTGGGGACATCATCACTGTGGCAGCTGGTCTGGATCGAGAGAAAGTGCAACAGTATACGTTAATAATTCAAGCCACAGACATGGAAGGCAATCCCACTTATGGCCTTTCAAACACAGCCACAGCCGTCATCACGGTGACAGATGTCAATGACAATCCTCCAGAGTTTACTGCCATGACTTTCTACGGAGAAGTCCCTGAGAACAGGGTGGACGTCAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M-R Lee et al.
Cell death & disease, 5, e1113-e1113 (2014-03-15)
Endoplasmic reticulum (ER) stress is considered one of the pathological mechanisms of idiopathic pulmonary fibrosis (IPF). Therefore, we examined whether an ER stress regulator, Bax inhibitor-1 (BI-1), regulates collagen accumulation, which is both a marker of fibrosis and a pathological
Yu-Huan Tsai et al.
PLoS pathogens, 9(5), e1003381-e1003381 (2013-06-06)
Listeria monocytogenes (Lm) is an invasive foodborne pathogen that leads to severe central nervous system and maternal-fetal infections. Lm ability to actively cross the intestinal barrier is one of its key pathogenic properties. Lm crosses the intestinal epithelium upon the
Maorong Jiang et al.
Neurochemical research, 39(11), 2047-2057 (2014-08-15)
Chitosan-based tissue engineered nerve grafts are successfully used for bridging peripheral nerve gaps. The biodegradation products of chitosan are water-dissolvable chitooligosaccharides (COSs), which have been shown to support peripheral nerve regeneration. In this study, we aimed to examine in vitro
Svetlana N Rubtsova et al.
PloS one, 10(7), e0133578-e0133578 (2015-07-25)
Using confocal microscopy, we analyzed the behavior of IAR-6-1, IAR1170, and IAR1162 transformed epithelial cells seeded onto the confluent monolayer of normal IAR-2 epithelial cells. Live-cell imaging of neoplastic cells stably expressing EGFP and of normal epithelial cells stably expressing
Xuebing Yan et al.
Molecular medicine reports, 12(2), 2999-3006 (2015-05-06)
Recent studies have indicated that the epithelial-mesenchymal transition (EMT) is a key molecular mechanism involved in the development of colorectal cancer (CRC). N-cadherin is a mesenchymal marker of the EMT and has been closely linked to several human malignancies. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service