Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | €212.00 |
50 μg | Please contact Customer Service for Availability | €382.00 |
About This Item
€212.00
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TGCTATGGGAGATTGGATGACTGTTACAGATCCAGGTCTGTCTTCAGAAAGCAAAACTATCTCTCAATATACCTCAGAAACAAAGATGTCTCCATCAAGTTTATACTCACAGCAAGTGCTATGTTCTTCAATACCTTTATCGAAAAATGTGCACAGTTTTTTCAGTGCCTTCTGCACAGAAGATAATATTGAACAGAGTATCTCATATCTTGATCAGGAATTGACTACTTTTGGTTTTCCTTCATTATATGAAGAATCCAAAGGTAAAGAGACAAAGAGAGAGTTAAATATAGTAGCTGTACTAAATTGTATGAATGAGCTGCTTGTGCTTCAGCGGAAGAACCTTCTAGCTCAGGAAAATGTGGAGACACAGAATTTGAAGCTGGGAAGTGATATGGACCATCTACAGAGCTGCTACTCAAAACTTAAGGAACAACTGGAAACCTCCAGGAGGGAAATGATTGGGCTTCAGGA
Ensembl | human accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... SSX2IP(117178)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| Gene Information human ... SSX2IP(117178) | Gene Information human ... DACH2(117154) | Gene Information human ... SAAL1(113174) | Gene Information human ... TADA1(117143), TADA1L(117143) |
| esiRNA cDNA target sequence TGCTATGGGAGATTGGATGACTGTTACAGATCCAGGTCTGTCTTCAGAAAGCAAAACTATCTCTCAATATACCTCAGAAACAAAGATGTCTCCATCAAGTTTATACTCACAGCAAGTGCTATGTTCTTCAATACCTTTATCGAAAAATGTGCACAGTTTTTTCAGTGCCTTCTGCACAGAAGATAATATTGAACAGAGTATCTCATATCTTGATCAGGAATTGACTACTTTTGGTTTTCCTTCATTATATGAAGAATCCAAAGGTAAAGAGACAAAGAGAGAGTTAAATATAGTAGCTGTACTAAATTGTATGAATGAGCTGCTTGTGCTTCAGCGGAAGAACCTTCTAGCTCAGGAAAATGTGGAGACACAGAATTTGAAGCTGGGAAGTGATATGGACCATCTACAGAGCTGCTACTCAAAACTTAAGGAACAACTGGAAACCTCCAGGAGGGAAATGATTGGGCTTCAGGA | esiRNA cDNA target sequence TATAACAGCTGCAGCGATGGCTGAGGCGATGAAACTTCAGAAGATGAAGCTTATGGCTATGAACACTCTTCAGGGAAATGGAAGCCAAAATGGGACCGAATCAGAGCCTGATGATCTTAATTCTAACACAGGTGGAAGTGAATCCTCCTGGGATAAAGATAAGATGCAGTCTCCATTTGCTGCACCTGGACCCCAACATGGAATTGCTCATGCAGCCCTAGCTGGCCAGCCAGGCATTGGGGGTGCTCCAACCCTCAATCCACTGCAGCAGAACCACCTGCTAACCAATAGACTGGATCTGCCATTTATGATGATGCCTCATCCCCTACTTCCAGTCAGCTTACCTCCTGCATCAGTTGCCATGGCAATGAATCAGATGAACCATCTCAATACTATTGCCAACATGGCTGC | esiRNA cDNA target sequence GGAGTACTGGCCAAGTCCAAGTGTCCTCGATTAAGAGAAATCTGTGTGGGAATTTTAGGTAATATGGCCTGTTTCCAGGAGATATGTGTGTCCATCAGCAGTGATAAAAATCTTGGGCAGGTGTTATTGCACTGTTTGTATGATTCAGACCCACCTACTCTGCTGGAAACAAGCAGGTTGTTGCTTACTTGCCTTTCCCAGGCAGAAGTGGCCAGTGTTTGGGTTGAAAGGATCCAGGAACATCCAGCTATTTATGATAGCATTTGCTTCATTATGTCAAGTTCAACAAATGTTGACTTGCTGGTGAAGGTGGGGGAGGTTGTGGACAAGCTCTTTGATTTGGATGAGAAACTAATGTTAGAATGGGTCAGAAATGGGGCTGCTCAGCCTCTGGACCAACCCCAGGAAGAGTCT | esiRNA cDNA target sequence TGGCATCCAAATCATGAAGAGCTGCAGCAAGACAAAGTTCACCGCCAGCGCTTGGCAGCCAAGGAGGGGCTTTTGCTGTGCTAAATTAGGATTTGAGGGTGTGGGACCCTCACCGAATTCATTGATTACTGAAAATTGAATGTTTTTTGGGTCCACATTTCAAGGCTGAAGTGTATAGTGTATATATAACCTTTCCTATGGAAATGTGACATTGAGTACATTTTGTGTTGCTGTTGTGAAGCCATTAATATAAATCTTTGGTAATGACCCATATCTCTATATGTATGTGTTCCCAGTTGTGGGAGCAGGCACTAATGAAATCCTGTGCCTGGAATGGAGATATTTAGGTACCTGAGGCTTAGTGTCCTGTGGTCTGCATGTAAGATAGATGACATCCTAGAACAAAGAAGCTGTTTTAACTTAATCCCCCTGATCAGCA |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
