Skip to Content
Merck
All Photos(1)

Documents

E4506

Sigma-Aldrich

Ethyl Violet

Synonym(s):

Basic violet 4, Ethyl purple 6B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C31H42N3Cl
CAS Number:
Molecular Weight:
492.14
Colour Index Number:
42600
Beilstein:
4117566
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, ~80%

SMILES string

[Cl-].CCN(CC)c1ccc(cc1)C(\c2ccc(cc2)N(CC)CC)=C3/C=C\C(C=C3)=[N+](\CC)CC

InChI

1S/C31H42N3.ClH/c1-7-32(8-2)28-19-13-25(14-20-28)31(26-15-21-29(22-16-26)33(9-3)10-4)27-17-23-30(24-18-27)34(11-5)12-6;/h13-24H,7-12H2,1-6H3;1H/q+1;/p-1

InChI key

JVICFMRAVNKDOE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service