DNB600
GenElute™ Plasma/Serum Cell-Free Circulating DNA Purification Midi Kit
About This Item
Recommended Products
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
shipped in
ambient
storage temp.
room temp
General description
- Isolate all sizes of circulating DNA from plasma and serum samples
- Isolate viral and bacterial DNA
- Versatile plasma and serum input volumes (1 mL – 4 mL)
- Concentrate circulating DNA into a flexible elution volume ranging from (50 μL - 100 μL)
- Isolate inhibitor-free cell-free circulating DNA
- Purify high-quality DNA in 40-45 minutes
- Compatible with Streck Cell-Free DNA BCT Tubes
Application
- PCR
- qPCR
- Southern Blot
- Methylation-sensitive PCR
- CpG array
- Restriction enzyme digestions
- Viral Detection
- Bacterial Detection
- Microarray
- NGS
Unit Definition
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service