Skip to Content
Merck

D8151

DIPSO sodium salt

≥99% (titration)

Synonym(s):

3-(N,N-Bis[2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C7H16NO6SNa
CAS Number:
Molecular Weight:
265.26
UNSPSC Code:
12161700
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C7H17NO6S.Na/c9-3-1-8(2-4-10)5-7(11)6-15(12,13)14;/h7,9-11H,1-6H2,(H,12,13,14);/q;+1/p-1

SMILES string

[Na+].OCCN(CCO)CC(O)CS([O-])(=O)=O

InChI key

UASJMJKAMKICKL-UHFFFAOYSA-M

assay

≥99% (titration)

form

crystalline powder

useful pH range

7.0-8.2

pKa (25 °C)

7.6

solubility

water: 0.333 g/L, clear to slightly hazy, colorless

application(s)

diagnostic assay manufacturing

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service