Skip to Content
Merck
All Photos(1)

Key Documents

D7644

Sigma-Aldrich

Disopyramide

Synonym(s):

α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29N3O
CAS Number:
Molecular Weight:
339.47
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC(C)N(CCC(C(N)=O)(c1ccccc1)c2ccccn2)C(C)C

InChI

1S/C21H29N3O/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25)

InChI key

UVTNFZQICZKOEM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Class IA antiarrhythmic; sodium channel blocker

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Y Kim et al.
Drug safety, 5(6), 393-420 (1990-11-01)
Quinidine, procainamide and disopyramide are antiarrhythmic drugs in the class 1A category. These drugs have a low toxic to therapeutic ratio, and their use is associated with a number of serious adverse effects during long term therapy and life-threatening sequelae
Ferdinando Pasquale et al.
Nature reviews. Cardiology, 6(4), 313-316 (2009-04-09)
A 61-year-old man presented with shortness of breath and chest pain on exertion. He had been diagnosed as having hiatus hernia 2 years previously and was taking proton-pump inhibitors as necessary. He had a family history of ischemic heart disease
B Brembilla-Perrot et al.
Archives des maladies du coeur et des vaisseaux, 84(6), 837-842 (1991-06-01)
The aim of this study was to assess the effect of delayed release Disopyramide 500 mg daily on sinus node function in 12 subjects with sinus node dysfunction and supraventricular and/or ventricular excitability. An ECG, Holter monitoring and electrophysiological studies
R N Brogden et al.
Drugs, 34(2), 151-187 (1987-08-01)
Disopyramide is a widely used class IA antiarrhythmic drug with a pharmacological profile of action similar to that of quinidine and procainamide. Over the past 10 years disopyramide has demonstrated its efficacy in ventricular and atrial arrhythmias. In therapeutic trials
J P Fauchier et al.
Archives des maladies du coeur et des vaisseaux, 84(4), 553-560 (1991-04-01)
Moricizine chlorhydrate (Ethmozine), a relatively unknown antiarrhythmic agent in France, is a derivative of Phenothiazine, related to the Vaughan-Williams Class IB drugs. A randomised, double-blind, crossover trial with Disopyramide 600 mg/day after a placebo period in 10 patients with ventricular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service