Skip to Content
Merck
All Photos(1)

Key Documents

D3939

Sigma-Aldrich

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride

peroxidase substrate, liquid, suitable for Immunohistology

Synonym(s):

DAB liquid substrate system

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride, for Immunohistology

Quality Level

form

liquid

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.Cl.Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4.4ClH/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;/h1-6H,13-16H2;4*1H

InChI key

KJDSORYAHBAGPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3′-Diaminobenzidine (DAB) is a chromogenic substrate for peroxidase enzyme. It is converted to an insoluble precipitate after oxidation by peroxidase.[1]

Application

3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride has been used:
  • in the detection of hydrogen peroxide in rice leaves[2]
  • as a substrate for rabbit peroxidase in immunohistochemistry of mice brain sections[3]
  • in immunocytochemistry of muscle-derived stem cells (MDSCs)[4]

The DAB (3,3′-Diaminobenzidine tetrahydrochloride) Two-component Enhanced Liquid Substrate System is a precipitating substrate system for peroxidase. DAB Two-component Liquid Substrate System produces a brown precipitate for use in immunohistological and immunoblotting staining procedures.

Caution

Stable for up to 18 mo. when stored at 2-8 °C in the original container.

Quantity

Each DAB Two-component Liquid Substrate System will provide 100 mL of the DAB substrate solution.

Kit Components Only

Product No.
Description

  • 3, 3′-Diaminobenzidine enhanced liquid substrate system, solution B, for immuno-histochemistry 100 mL

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of salinity tolerance in rice by constitutive-overexpression of genes involved in the regulation of programmed cell death
Hoang TML
Frontiers in Plant Science, 6, 175-175 (2015)
The neuroprotective effects of muscle-derived stem cells via brain-derived neurotrophic factor in spinal cord injury model
Han D, et al.
BioMed Research International, 2017 (2017)
Chuangao Xie et al.
Oncology letters, 16(1), 940-948 (2018-07-03)
The purpose of the present study was to elucidate the effects of cyclooxygenase 2 (COX-2) on the expression of vascular endothelial growth factor (VEGF) and prostaglandin E2 (PGE2) in pancreatic cancer in vitro and in vivo, and to clarify the
Mingzhu Yin et al.
Cell reports, 27(9), 2709-2724 (2019-05-30)
The regenerative capacity of the human endometrium requires a population of local stem cells. However, the phenotypes, locations, and origin of these cells are still unknown. In a mouse menstruation model, uterine stromal SM22α+-derived CD34+KLF4+ stem cells are activated and
Near-infrared light (670 nm) reduces MPTP-induced parkinsonism within a broad therapeutic time window
Reinhart F, et al.
Experimental Brain Research. Experimentelle Hirnforschung. Experimentation Cerebrale, 234(7), 1787-1794 (2016)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service