Skip to Content
Merck

Skip To

D1071

Decoyinine

Synonym(s):

9-(6-Deoxy-D-β-erythro-hex-5-en-2-ulofuranosyl)adenine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H13N5O4
CAS Number:
Molecular Weight:
279.25
PubChem Substance ID:
UNSPSC Code:
51101500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

antibiotic activity spectrum

Gram-positive bacteria

mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(cnc12)[C@]3(CO)OC(=C)[C@@H](O)[C@H]3O

InChI

1S/C11H13N5O4/c1-5-7(18)8(19)11(2-17,20-5)16-4-15-6-9(12)13-3-14-10(6)16/h3-4,7-8,17-19H,1-2H2,(H2,12,13,14)/t7-,8-,11-/m1/s1

InChI key

UZSSGAOAYPICBZ-SOCHQFKDSA-N

Application

Decoyinine is used to study the inhibition of cell wall synthesis, to inhibit RNA synthesis, to lower intracellular GTP levels in Streptococcus mutans, and to initiate sporulation in Bacillus.

Biochem/physiol Actions

Decoyinine is a Xanthosine monophosphate (XMP) aminase inhibitor and GMP synthetase inhibitor.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library