Skip to Content
Merck
All Photos(1)

Documents

CLS3833

Corning® CellBIND® 1536 well microplates

black, clear bottom, low base, smooth top, w/o lid, sterile

Synonym(s):

1536 multiwell plates, 1536 well microplates, 1536 well microtiter plates, 1536 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

clear wells
polystyrene plate
square wells

sterility

sterile

feature

lid: no
black, clear bottom

packaging

case of 100 ea

manufacturer/tradename

Corning 3833

size

1536 wells

well volume

12.5 μL

well working volume

1-8 μL

color

black

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service