Skip to Content
Merck
All Photos(1)

Key Documents

CLS356468

Corning® BioCoat Poly-D-Lysine Culture Dish

diam. 60 mm

Synonym(s):

Cell Culture Dish, plastic cell culture dish, coated cell culture dish

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
€530.00

€530.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EACH
€530.00

About This Item

UNSPSC Code:
41121812
NACRES:
NB.13

€530.00


Please contact Customer Service for Availability

feature

lid

packaging

pkg of 100 ea (5 packs)

manufacturer/tradename

Corning 356468

diam.

60 mm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service