Skip to Content
Merck
All Photos(2)

Key Documents

CLS354632

Corning® BioCoat Poly-D-Lysine Culture Slide

capacity 8 well

Synonym(s):

slides, chambers

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 12 EACH
€482.00

€482.00


Please contact Customer Service for Availability


Select a Size

Change View
3 X 12 EACH
€482.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

€482.00


Please contact Customer Service for Availability

packaging

pkg of 12 (3 packs )

manufacturer/tradename

Corning 354632

capacity

8 well

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service