CLS354253
Corning® Cell Recovery Solution
pack of 100 mL
Synonym(s):
Cell Recovery Medium, Recovery Solution
Select a Size
€102.00
In StockDetails
Select a Size
About This Item
€102.00
In StockDetails
Recommended Products
packaging
pack of 100 mL
manufacturer/tradename
Corning 354253
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Quality
Tested for ability to depolymerize a 1mm thick layer of gelled Matrigel Matrix after one hour at 2° to 8°C
Filtered (0.2 μm membrane) and tested for bacteria, fungi, and mycoplasma
Preparation and Storage
Stable for at least three months at 2° to 8°C.
Handling
Corning Cell Recovery Solution is used full strength to recover cells from Corning Matrigel Matrix.
Formulation
Nonenzymatic proprietary solution
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service