Skip to Content
Merck
All Photos(2)

Key Documents

CLS353096

Corning® Falcon® Permeable Support

for use with 24 Well Plate, with 3 µm Transparent PET Membrane, sterile

Synonym(s):

Permeable Support

Sign Into View Organizational & Contract Pricing

Select a Size

48 EACH
€435.00

€435.00


Please contact Customer Service for Availability


Select a Size

Change View
48 EACH
€435.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

€435.00


Please contact Customer Service for Availability

sterility

sterile

feature

with 3 µm Transparent PET Membrane

packaging

case of 48 ea (Individually wrapped)

manufacturer/tradename

Corning 353096

compatibility

for use with 24 Well Plate

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® Permeable Support for 24 Well Plate with 3.0µm Transparent Polyester (PET) Membrane, Sterile, 1/Pack, 48/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service