CLLS1006
SW48 Cells BAX -/-
Human female colon, colorectal adenocarcinoma
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
General description
SW48 Cells BAX -/- are colorectal adenocarcinoma, epithelial cells, from a human caucasian female (aged 82 years), with a ZFN knock out modification.
This product corresponds to ATCC Cat. No. CCL-231
Application
Media Renewal changes one to two times per week
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:2 to 1:6
The base medium for this cell line is DMEM, Cat. No. D5671. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, Sodium pyruvate, Cat. No. S8636, at a final concentration of 1 mM, and L-glutamine, Cat. No. G7513, at a final concentration of 2mM.
The cell freezing medium is DMSO 1X (Cat. No. C6164)
Features and Benefits
These SW48 cells are adherent, with a doubling time of approx. 35 hours.
Zinc finger nuclease (ZFN) knock out on chromosome 19q13.3-13.4
Quality
Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service