Skip to Content
Merck
All Photos(1)

Documents

PE/CA-PJ41 (clone D2)

98020207, human oral squamous epithelium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PE/CA-PJ41 (clone D2), 98020207

biological source

human mouth (oral)

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human oral squamous cell carcinoma

Cell Line Description

PE/CA-PJ41 (clone D2) has been derived from the oral squamous epithelium of a 67 year old female patient. The cell line, also known as Plaepi 41, has been shown to express cytokeratin.

Application

Cancer Studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 11,12
D16S539: 11,13
D5S818: 12
D7S820: 9,11
THO1: 7
TPOX: 8
vWA: 15,19

Culture Medium

IDMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:8 i.e. seeding at 1-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service