Skip to Content
Merck
All Photos(1)

Key Documents

SK-PN-DW

94092301, human nerve, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SK-PN-DW, 94092301

biological source

human nerve

growth mode

Semi-adherent

karyotype

82XXY

morphology

Polygonal

products

Pro-cholecystokinin (Pro-cck)

receptors

Vimentin, P53, anti-MIC2

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human primitive neuroectodermal tumour

Cell Line Description

SK-PN-DW has been derived from a male patient with a primitive neuroectodermal tumour that expresses cholecystokinin (cck) RNA and synthesises substantial amounts of pro-cck. SK-PN-DW is able to process the pro-hormone, producing immunoreactive cck-like material plus a peptide that immunochemically and chromatographically resembles intact cck c-terminal extension peptide. Cells have a tendency to float.

Application

Tumour lines should prove useful in understanding the regulation of the cck gene in human neuronal cells and will also provide an in vitro system for investigation of the post translational processing of the cck preprohormone.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 9
D16S539: 12
D5S818: 13,14
D7S820: 8,14
THO1: 6
TPOX: 8,9
vWA: 16

Culture Medium

DMEM or EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service