Skip to Content
Merck
All Photos(1)

Key Documents

MEG-01

94012401, human bone marrow, Myoblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MEG-01, 94012401

biological source

human bone marrow

growth mode

Suspension

karyotype

Modal no. 56-58, hyperdiploid

morphology

Myoblast-like

products

Immunoglobulin G (IgG)-Fc in 12% of cells; C3′ in 53% of cells

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human megakaryoblastic leukaemia

Cell Line Description

Derived from the bone marrow of a 55 year old male with chronic myelogenous leukaemia. All karyotypes show the Philadelphia chromosome. Platelet GP IIb/IIIa antigen is expressed on more than 92% of cells. Cytoplasmic expression of GP Ib antigen was not detected on the surface of cells but was present in the cytoplasm 20% of cells. BA-1 antigen is expressed in almost all cells. FVIII-related antigen was not detected on the surface cells but was present in cytoplasm.

Application

Studies of physiological mechanisms governing the differentiation of human megakaryocyte and platelet release; the study of the biochemistry (and differentiation) of erythroid and myeloid cells.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10
D13S317: 8
D16S539: 9
D5S818: 13
D7S820: 11
THO1: 7
TPOX: 8,11
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Subcultures are prepared by diluting the cell suspension into fresh medium at a ratio of 1:2 to 1:3. Cells should be seeded at 2-4 x 105 cells/ml. Doubling time is 36-48 hours. 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service