Skip to Content
Merck
All Photos(1)

Documents

A2058

91100402, human skin (lymph node metastasis), Epithelial

Synonym(s):

A2058 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

A2058, 91100402

biological source

human skin (lymph node metastasis)

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Nerve growth factor (NGF), Laminin

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human caucasian metastatic melanoma

Cell Line Description

This cell line has been isolated from a lymph node metastasis from a 43 year old male patient with malignant melanoma. Due to their highly invasive properties, the cells can be used as a source of cellular invasion associated proteins e.g. collagen type IV collagenase, tissue inhibitor of metalloproteinase-2 and autocrine motility factor.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service