Skip to Content
Merck
All Photos(1)

Key Documents

DHD/K12/TRb

90062901, rat colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
rat colon
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer
Pricing and availability is not currently available.

Product Name

DHD/K12/TRb, 90062901

biological source

rat colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat colonic carcinoma

Cell Line Description

This cell line has properties essential for study of metastasis of colon carcinomas; a reproducible pattern of metastatic behaviour and ability to be propagated both in vitro and in vivo to facilitate identification of biochemical correlates of metastasis.

Application

Metastasis studies

Culture Medium

DMEM + F10 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) or 2% Rat Serum.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service