Skip to Content
Merck
All Photos(1)

Documents

WEHI-231

85022107, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

WEHI-231, 85022107

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Surface Immunoglobulin M (IgM)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse B cell lymphoma

Cell Line Description

Expresses surface IgM but does not secrete Ig. Secretion of IgM can be induced with LPS. WEHI 231 is a B cell lymphoma of BALB/c x NXB F1 origin induced by mineral oil injection.

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 to 1:10 i.e. seeding at 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service