Skip to Content
Merck
All Photos(1)

Documents

NS20Y-BU-4

8062503, mouse nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NS20Y-BU-4, 08062503

biological source

mouse nerve

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Choline acetyltransferase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma

Cell Line Description

A bromodeoxyuridine-resistant mutant of NS20Y (Sigma Catalogue number 08062517) The cell line NS20Y is a cholinergic cell line derived from mouse neuroblastoma C1300 Mouse strain A/Jax.

Application

Used in the generation of cell hybrids.

Culture Medium

DMEM + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS)+ 1 mM 8-bromodeoxyuridine. Cells can also be grown in the absence of bromodeoxyuridine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service