Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~19 kDa
species reactivity
human
technique(s)
western blot: 1:500-1:2,000
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CIRBP(1153)
General description
CIRBP is expressed in normal testes and downregulated after heat stress caused by cryptorchidism, varicocele or environmental temperatures.
Immunogen
synthetic peptide corresponding to amino acids 121-135 of human CIRBP
Application
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if. Anti-CIRBP antibody produced in rabbit is suitable for western blotting at a working dilution of 1:500-1:2000.
Biochem/physiol Actions
Extracellular CIRBP is a detrimental factor in stimulating inflammation to cause neuronal damage in cerebral ischemia.[1] It may have a protective effect on testicular injury after testicular torsion/detorsion when upregulated. Downregulation of CIRBP has been shown to increase germ cell apoptosis.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.