Skip to Content
Merck
All Photos(1)

Key Documents

C9005

Sigma-Aldrich

L-Cystine-di-2-naphthylamide

Synonym(s):

L-Cystine di-β-naphthylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N4O2S2
CAS Number:
Molecular Weight:
490.64
Beilstein:
3183259
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, light yellow to yellow

storage temp.

−20°C

SMILES string

N[C@@H](CSSC[C@H](N)C(=O)Nc1ccc2ccccc2c1)C(=O)Nc3ccc4ccccc4c3

InChI

1S/C26H26N4O2S2/c27-23(25(31)29-21-11-9-17-5-1-3-7-19(17)13-21)15-33-34-16-24(28)26(32)30-22-12-10-18-6-2-4-8-20(18)14-22/h1-14,23-24H,15-16,27-28H2,(H,29,31)(H,30,32)/t23-,24-/m0/s1

InChI key

REEVAJUPLLWYBH-ZEQRLZLVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P F Silveira et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 102(1), 119-122 (1992-05-01)
1. Bothrops jararaca plasma or serum hydrolysed L-cystine-di-beta-naphthylamide (CNAse activity) at a degree comparable to that of plasma or serum in pregnant women. 2. In adult snakes, activity was less in males. It was not altered in pregnancy but increased
[Evaluation of substrates for measuring cystyl-amino peptidase activity in various sera and tissues with respect to L-cystine-di-beta-naphthylamide, L-cystine-bis-p-nitroanilide, L-cysteine-beta-naphthylamide, and S-benzyl-L-cysteine-p-nitroanilide (author's transl)].
H Motokura et al.
Rinsho byori. The Japanese journal of clinical pathology, 28(4), 393-397 (1980-04-01)
Comparison of substrates for measuring cystyl-amino peptidase activity in serum of pregnancy and hepatic disease and in various tissues.
T Uete et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 19(3), 145-154 (1981-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service